(4S,5R)-2-(6-Cyclopropylpyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole ≥97%

Supplier: Ambeed
Warning

A1519878-250MG A1519878-1G
77597-918EA 271.63 USD
77597-918 77625-052
(4S,5R)-2-(6-Cyclopropylpyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole ≥97%
(4S,5R)-2-(6-Cyclopropylpyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₃H₂₀N₂O
MW: 340.41 g/mol
CAS Number: 2828438-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR