(S)-2,2'-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)-1,1'-binaphthalene ≥97%

Supplier: Ambeed
Warning

A1519834-100MG A1519834-1G A1519834-250MG
77624-984EA 108.15 USD
77624-984 77624-986 77624-988
(S)-2,2'-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)-1,1'-binaphthalene ≥97%
(S)-2,2'-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)-1,1'-binaphthalene

Precaución: for Research Use Only

Formula: C₄₀H₃₂N₂O₂
MW: 572.69 g/mol
CAS Number: 1603007-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR