(S)-2,2'-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)-1,1'-binaphthalene ≥97%
Supplier: Ambeed
|
Warning
|
A1519832-1G
A1519832-250MG
A1519832-100MG
77624-980EA
619.03
USD
77624-980
77624-982
77624-978
(S)-2,2'-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)-1,1'-binaphthalene ≥97%
(S)-2,2'-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)-1,1'-binaphthalene
Precaución: for Research Use Only
Formula:
C₄₀H₃₂N₂O₂ MW: 572.69 g/mol |
CAS Number:
180857-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...