(R)-2-(6-Cyclopropylpyridin-2-yl)-4-phenyl-4,5-dihydrooxazole ≥95%

Supplier: Ambeed
Warning

A1519817-250MG A1519817-1G
77597-912EA 164 USD
77597-912 77624-942
(R)-2-(6-Cyclopropylpyridin-2-yl)-4-phenyl-4,5-dihydrooxazole ≥95%
(R)-2-(6-Cyclopropylpyridin-2-yl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂O
MW: 264.32 g/mol
CAS Number: 2828438-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR