Hyaluronic acid sodium salt, M.W. 500000-750000 Da

Supplier: Ambeed
Warning

Synonyms: Sodium hyaluronate, Hyaluronic acid sodium, HA sodium salt

A1519704-1G A1519704-100G A1519704-5G
77597-908EA 6.15 USD
77597-908 77624-894 77624-896
Hyaluronic acid sodium salt, M.W. 500000-750000 Da
Hyaluronic acid sodium salt

Precaución: for Research Use Only

Formula: (C₁₄H₂₁NaNO₁₁)n
MDL Number: MFCD00875848
CAS Number: 9067-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR