(4S,5S)-1,3-Bis(2-isopropoxyphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate ≥97%
Supplier: Ambeed
|
Danger
|
A1517145-1G
A1517145-250MG
77876-000EA
860
USD
77876-000
77876-001
(4S,5S)-1,3-Bis(2-isopropoxyphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate ≥97%
(4S,5S)-1,3-Bis(2-isopropoxyphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate
Precaución: for Research Use Only
CAS Number:
1033618-51-7 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...