Pyren-2-ylboronic acid ≥98%

Supplier: Ambeed
Warning

A1517106-100MG A1517106-1G A1517106-250MG
77875-995EA 91 USD
77875-995 77875-996 77875-997
Pyren-2-ylboronic acid ≥98%
Pyren-2-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₁BO₂
CAS Number: 1195975-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR