Ethyl 4-hydroxy-2-methylbenzoate ≥97%

Supplier: Ambeed
Warning

A151683-5G A151683-25G
77615-226EA 49.44 USD
77615-226 77746-048
Ethyl 4-hydroxy-2-methylbenzoate ≥97%
Ethyl 4-hydroxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
MDL Number: MFCD12025348
CAS Number: 57081-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR