[[2,2'-[(1S,2S)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)-6-methyl-phenolato-κO]](2-)][tetrafluoroborato(1-)-κF]cobalt ≥97%

Supplier: Ambeed
Danger

A1516759-100MG A1516759-250MG
77597-894EA 56.38 USD
77597-894 77597-896
[[2,2'-[(1S,2S)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)-6-methyl-phenolato-κO]](2-)][tetrafluoroborato(1-)-κF]cobalt ≥97%
[[2,2'-[(1S,2S)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)-6-methyl-phenolato-κO]](2-)][tetrafluoroborato(1-)-κF]cobalt

Precaución: for Research Use Only

Formula: C₃₀H₄₀BCoF₄N₂O₂
MW: 606.3886 g/mol
CAS Number: 2828438-37-3
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR