[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-(1,1-dimethylethyl)-6-methyl-phenolato-κO]](2-)](nitrato-κO)cobalt ≥97%

Supplier: Ambeed
Danger

A1516754-100MG A1516754-250MG A1516754-1G
77597-890EA 54.33 USD
77597-890 77597-892 77624-860
[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-(1,1-dimethylethyl)-6-methyl-phenolato-κO]](2-)](nitrato-κO)cobalt ≥97%
[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-(1,1-dimethylethyl)-6-methyl-phenolato-κO]](2-)](nitrato-κO)cobalt

Precaución: for Research Use Only

Formula: C₃₀H₄₀CoN₃O₅
MW: 581.58 g/mol
CAS Number: 2828438-32-8
UN: 1479
ADR: 5.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR