(4R,5R)-1,3-Dimesityl-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate ≥98%
Supplier: Ambeed
|
Danger
|
A1515926-100MG
A1515926-1G
A1515926-250MG
77624-848EA
202.91
USD
77624-848
77875-983
77875-984
(4R,5R)-1,3-Dimesityl-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate ≥98%
(4R,5R)-1,3-Dimesityl-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₃₃H₃₅BF₄N₂ |
CAS Number:
372517-11-8 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...