Boc-L-Orn(Aloc)-OH 98+%

Supplier: Ambeed
Warning

Synonyms: Boc-Orn(Aloc)-OH, Aloc-Orn(Boc)-OH · DCHA, (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(prop-2-enoxycarbonylamino)pentanoic acid

A151575-25G A151575-5G
77900-968EA 124 USD
77900-968 77901-695
Boc-L-Orn(Aloc)-OH 98+%
Boc-L-Orn(Aloc)-OH

Precaución: for Research Use Only

Formula: C₁₄H₂₅N₂O₆
MW: 316.4 g/mol
MDL Number: MFCD00800349
CAS Number: 171820-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR