(OC-6-42)-Bis[[2,3-butanedione 2,3-di(oximato-κN)](1-)]chloro(4-methoxypyridine-κN)cobalt ≥97%

Supplier: Ambeed
Warning

A1515462-100MG A1515462-1G A1515462-250MG
77597-872EA 35.88 USD
77597-872 77597-874 77624-844
(OC-6-42)-Bis[[2,3-butanedione 2,3-di(oximato-κN)](1-)]chloro(4-methoxypyridine-κN)cobalt ≥97%
(OC-6-42)-Bis[[2,3-butanedione 2,3-di(oximato-κN)](1-)]chloro(4-methoxypyridine-κN)cobalt

Precaución: for Research Use Only

Formula: C₁₄H₂₁ClCoN₅O₅
MW: 433.73 g/mol
CAS Number: 1574577-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR