Polyethylene glycol 3,400

Supplier: Ambeed
Warning

Synonyms: Ethylene glycol polymer 3350, PEG 3,350, PEG 3400

A1515450-100G A1515450-500G
77597-868EA 7.18 USD
77597-868 77597-870
Polyethylene glycol 3,400
Polyethylene glycol 3,400

Precaución: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MW: 3400 g/mol
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR