Polyethylene glycol dimethylether Mn ~250

Supplier: Ambeed
Warning

Synonyms: Poly(ethylene glycol) dimethyl ether Mn 250

A1515436-100G A1515436-500G
77624-836EA 20.6 USD
77624-836 77624-838
Polyethylene glycol dimethylether Mn ~250
Polyethylenglycoldimethylether

Precaución: for Research Use Only

Formula: H₃C(OCH₂CH₂)nOCH₃
MDL Number: MFCD00147972
CAS Number: 24991-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR