Methyl 2-methoxy-3-nitrobenzoate ≥98%

Supplier: Ambeed
Warning

A151492-1G A151492-5G
77889-994EA 23 USD
77889-994 77889-995
Methyl 2-methoxy-3-nitrobenzoate ≥98%
Methyl 2-methoxy-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MDL Number: MFCD11045625
CAS Number: 90564-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR