Ethyl 3-(2,5-difluorophenyl)-3-oxopropanoate ≥97%

Supplier: Ambeed
Warning

A151386-1G A151386-5G
77879-051EA 17 USD
77879-051 77879-052
Ethyl 3-(2,5-difluorophenyl)-3-oxopropanoate ≥97%
Ethyl 3-(2,5-difluorophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₂O₃
MDL Number: MFCD07783808
CAS Number: 887267-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR