(R,E)-Cyclooct-4-en-1-yl (4-nitrophenyl) carbonate 98+% rel-(1R-4E-pR)-equatorial

Supplier: Ambeed
Warning

Synonyms: TCO-pnb ester

A1513369-100MG A1513369-250MG A1513369-50MG
77597-858EA 486.88 USD
77597-858 77597-860 77597-862
(R,E)-Cyclooct-4-en-1-yl (4-nitrophenyl) carbonate 98+% rel-(1R-4E-pR)-equatorial
(R,E)-Cyclooct-4-en-1-yl (4-nitrophenyl) carbonate

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO₅
MW: 291.3 g/mol
MDL Number: MFCD29052196
CAS Number: 1438415-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR