Ethyl 4-(aminomethyl)benzoate ≥95%
Supplier: Ambeed
|
Warning
|
A151304-1G
A151304-250MG
77611-412EA
46.35
USD
77611-412
77683-276
Ethyl 4-(aminomethyl)benzoate ≥95%
Ethyl 4-(aminomethyl)benzoate
Precaución: For research use only.
Formula:
C₁₀H₁₃NO₂ MW: 179.22 g/mol |
MDL Number:
MFCD00748342 CAS Number: 366-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...