4-Bromo-3-chlorophenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (4-Bromo-3-chlorophenyl)boronic acid

A151281-250MG A151281-5G
77883-188EA 19 USD
77883-188 77883-189
4-Bromo-3-chlorophenylboronic acid ≥98%
4-Bromo-3-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrClO₂
MDL Number: MFCD12913945
CAS Number: 1217501-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR