2,2',7,7'-Tetrakis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[fluorene] ≥95%
Supplier: Ambeed
|
Warning
|
A1512486-100MG
A1512486-1G
A1512486-250MG
A1512486-5G
77875-956EA
70
USD
77875-956
77875-957
77875-958
77875-959
2,2',7,7'-Tetrakis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[fluorene] ≥95%
2,2',7,7'-Tetrakis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[fluorene]
Precaución: for Research Use Only
Formula:
C₄₉H₆₀B₄O₈ |
CAS Number:
934391-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...