(3S,4R)-3-Fluoro-1-methylpiperidin-4-amine dihydrochloride ≥95%

Supplier: Ambeed
Warning

A1510243-250MG A1510243-5G A1510243-100MG
77597-844EA 60.47 USD
77597-844 77597-846 77875-924
(3S,4R)-3-Fluoro-1-methylpiperidin-4-amine dihydrochloride ≥95%
(3S,4R)-3-Fluoro-1-methylpiperidin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₅Cl₂FN₂
MDL Number: MFCD31743843
CAS Number: 2306246-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR