1-Methylcyclobutane-1,3-diol ≥95%

Supplier: Ambeed
Warning

A1509530-250MG A1509530-100MG A1509530-1G A1509530-5G
77597-842EA 235.75 USD
77597-842 77624-804 77624-806 77875-923
1-Methylcyclobutane-1,3-diol ≥95%
1-Methylcyclobutane-1,3-diol

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MDL Number: MFCD33404933
CAS Number: 2168552-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR