(R)-5-(8,9,10,11,12,13,14,15-Octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-5H-dibenzo[b,f]azepine ≥95%

Supplier: Ambeed
Warning

A1509185-100MG A1509185-1G A1509185-250MG
77597-836EA 116.85 USD
77597-836 77597-838 77597-840
(R)-5-(8,9,10,11,12,13,14,15-Octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-5H-dibenzo[b,f]azepine ≥95%
(R)-5-(8,9,10,11,12,13,14,15-Octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-5H-dibenzo[b,f]azepine

Precaución: for Research Use Only

Formula: C₃₄H₃₀NO₂P
MW: 515.5813 g/mol
CAS Number: 2070926-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR