(+)-Dibenzoyl-D-tartaric acid monohydrate ≥99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,3S)-2,3-Bis(benzoyloxy)succinic acid hydrate
A1509141-500G
A1509141-1000G
77597-834EA
39.98
USD
77597-834
77875-919
(+)-Dibenzoyl-D-tartaric acid monohydrate ≥99%
(+)-Dibenzoyl-D-tartaric acid monohydrate
Precaución: for Research Use Only
Formula:
C₁₈H₁₄O₈·1H₂O |
MDL Number:
MFCD00150722 CAS Number: 80822-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...