(+)-Dibenzoyl-D-tartaric acid ≥99%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-(+)-Di-O-benzoyltartaric acid, (2S,3S)-2,3-Bis(benzoyloxy)succinic acid

A1509139-100G A1509139-500G
77624-798EA 20.6 USD
77624-798 77875-918
(+)-Dibenzoyl-D-tartaric acid ≥99%
(+)-Dibenzoyl-D-tartaric acid

Precaución: for Research Use Only

Formula: C₁₈H₁₄O₈
MDL Number: MFCD00063222
CAS Number: 17026-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR