1-(2-Bromo-4-(trifluoromethyl)phenyl)ethanone ≥98%

Supplier: Ambeed
Warning

A150836-1G A150836-250MG A150836-5G
77892-028EA 40 USD
77892-028 77892-029 77892-030
1-(2-Bromo-4-(trifluoromethyl)phenyl)ethanone ≥98%
1-(2-Bromo-4-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O
MDL Number: MFCD11865317
CAS Number: 1131605-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR