3-(1-Oxo-5-(piperazin-1-yl)isoindolin-2-yl)piperidine-2,6-dione hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: Lenalidomide 5'-piperazine

A1508318-1G A1508318-100MG A1508318-250MG A1508318-10G
77597-828EA 234.73 USD
77597-828 77624-794 77624-796 77875-907
3-(1-Oxo-5-(piperazin-1-yl)isoindolin-2-yl)piperidine-2,6-dione hydrochloride ≥95%
3-(1-Oxo-5-(piperazin-1-yl)isoindolin-2-yl)piperidine-2,6-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₁ClN₄O₃
MW: 364.83 g/mol
MDL Number: MFCD32705051
CAS Number: 2222120-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR