3,5-Dihydro-1H-furo[3,4-c]pyrrole-4-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A1508141-1G A1508141-250MG
77597-820EA 175.28 USD
77597-820 77624-782
3,5-Dihydro-1H-furo[3,4-c]pyrrole-4-carboxylic acid ≥95%
3,5-Dihydro-1H-furo[3,4-c]pyrrole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.1354 g/mol
MDL Number: MFCD12923946
CAS Number: 535170-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR