Iridium-(4,4'-dimethoxy-2,2'-bipyridine-κN1,κN1')bis[(3-fluoro-5-trifluoromethyl)-2-(5-trifluoromethyl-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate ≥97%

Supplier: Ambeed
Warning

A1507468-100MG A1507468-250MG A1507468-50MG A1507468-1G
77624-768EA 290.46 USD
77624-768 77624-770 77624-772 77875-892
Iridium-(4,4'-dimethoxy-2,2'-bipyridine-κN1,κN1')bis[(3-fluoro-5-trifluoromethyl)-2-(5-trifluoromethyl-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate ≥97%
Iridium-(4,4'-dimethoxy-2,2'-bipyridine-κN1,κN1')bis[(3-fluoro-5-trifluoromethyl)-2-(5-trifluoromethyl-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate

Precaución: for Research Use Only

CAS Number: 2828438-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR