Iridium-(1,10-phenanthroline-κN1,κN10)bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate ≥95%

Supplier: Ambeed
Warning

A1507446-250MG A1507446-100MG A1507446-1G A1507446-50MG
77597-816EA 370.03 USD
77597-816 77624-754 77875-884 77875-885
Iridium-(1,10-phenanthroline-κN1,κN10)bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate ≥95%
Iridium-(1,10-phenanthroline-κN1,κN10)bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₄H₁₈F₁₂IrN₄P
CAS Number: 2828437-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR