6,7-Difluoro-1H-indole ≥95%

Supplier: Ambeed
Warning

A150603-1G A150603-250MG
77889-988EA 520 USD
77889-988 77889-989
6,7-Difluoro-1H-indole ≥95%
6,7-Difluoro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅F₂N
MDL Number: MFCD03095185
CAS Number: 271780-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR