4-((2-((Furan-2-ylmethyl)amino)-1H-benzo[d]imidazol-1-yl)methyl)benzonitrile 98+%

Supplier: Ambeed
Warning

Synonyms: TRPC5-IN-1

A1505669-100MG A1505669-1G A1505669-250MG
77875-872EA 70 USD
77875-872 77875-873 77875-874
4-((2-((Furan-2-ylmethyl)amino)-1H-benzo[d]imidazol-1-yl)methyl)benzonitrile 98+%
4-((2-((Furan-2-ylmethyl)amino)-1H-benzo[d]imidazol-1-yl)methyl)benzonitrile

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₄O
MW: 328.37 g/mol
CAS Number: 2265215-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR