tert-Butyl 3-(2,7-dichloro-8-fluoropyrido[4,3-d]pyrimidin-4-yl)-3,8-diazabicyclo[3.2.1]octane-8-carboxylate ≥97%

Supplier: Ambeed
Warning

A1505409-100MG A1505409-1G A1505409-250MG A1505409-5G
77875-866EA 97 USD
77875-866 77875-867 77875-868 77875-869
tert-Butyl 3-(2,7-dichloro-8-fluoropyrido[4,3-d]pyrimidin-4-yl)-3,8-diazabicyclo[3.2.1]octane-8-carboxylate ≥97%
tert-Butyl 3-(2,7-dichloro-8-fluoropyrido[4,3-d]pyrimidin-4-yl)-3,8-diazabicyclo[3.2.1]octane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₀Cl₂FN₅O₂
MDL Number: MFCD34471585
CAS Number: 2454397-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR