2,5-Dioxopyrrolidin-1-yl 3-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido)propanoate ≥95%

Supplier: Ambeed
Warning

A1504899-250MG A1504899-100MG
77597-808EA 112.75 USD
77597-808 77624-742
2,5-Dioxopyrrolidin-1-yl 3-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido)propanoate ≥95%
2,5-Dioxopyrrolidin-1-yl 3-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido)propanoate

Precaución: for Research Use Only

Formula: C₁₄H₁₅N₃O₇
MW: 337.28 g/mol
CAS Number: 2512227-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR