3-(3-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-3-oxopropoxy)propanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: VH 032 amide-PEG1-acid

A1504747-50MG A1504747-100MG A1504747-250MG A1504747-1G
77600-132EA 132.23 USD
77600-132 77597-804 77597-806 77624-738
3-(3-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-3-oxopropoxy)propanoic acid ≥95%
3-(3-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-3-oxopropoxy)propanoic acid

Precaución: for Research Use Only

Formula: C₂₈H₃₈N₄O₇S
MW: 574.69 g/mol
CAS Number: 2172820-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR