3-((4-(Piperidin-4-yl)phenyl)amino)piperidine-2,6-dione hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-dione hydrochloride"
A1503667-250MG
A1503667-100MG
77597-798EA
107.63
USD
77597-798
77624-728
3-((4-(Piperidin-4-yl)phenyl)amino)piperidine-2,6-dione hydrochloride ≥98%
3-((4-(Piperidin-4-yl)phenyl)amino)piperidine-2,6-dione hydrochloride
Precaución: for Research Use Only
Formula:
C₁₆H₂₂ClN₃O₂ MW: 323.82 g/mol |
CAS Number:
2259851-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...