3-((4-(Piperazin-1-yl)phenyl)amino)piperidine-2,6-dione hydrochloride ≥95%

Supplier: Ambeed
Warning

A1503666-100MG A1503666-250MG
77597-794EA 104.55 USD
77597-794 77597-796
3-((4-(Piperazin-1-yl)phenyl)amino)piperidine-2,6-dione hydrochloride ≥95%
3-((4-(Piperazin-1-yl)phenyl)amino)piperidine-2,6-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₁ClN₄O₂
MW: 324.81 g/mol
CAS Number: 2259851-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR