5,7-Di-tert-butyl-3-(3-fluorophenyl)benzo[d]oxazol-3-ium tetrafluoroborate ≥97%

Supplier: Ambeed
Danger

A1503655-250MG A1503655-100MG
77592-828EA 154.78 USD
77592-828 77624-726
5,7-Di-tert-butyl-3-(3-fluorophenyl)benzo[d]oxazol-3-ium tetrafluoroborate ≥97%
5,7-Di-tert-butyl-3-(3-fluorophenyl)benzo[d]oxazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₁H₂₅BF₅NO
MW: 413.23 g/mol
CAS Number: 2750161-96-5
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR