5,7-Di-tert-butyl-3-(4-(trifluoromethoxy)phenyl)benzo[d]oxazol-3-ium tetrafluoroborate ≥97%

Supplier: Ambeed
Danger

A1503652-250MG A1503652-100MG
77597-784EA 98.4 USD
77597-784 77597-782
5,7-Di-tert-butyl-3-(4-(trifluoromethoxy)phenyl)benzo[d]oxazol-3-ium tetrafluoroborate ≥97%
5,7-Di-tert-butyl-3-(4-(trifluoromethoxy)phenyl)benzo[d]oxazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₂H₂₅BF₇NO₂
MW: 479.2392 g/mol
CAS Number: 2750161-90-9
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR