2-(7,8-Difluoro-3-(methoxymethoxy)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
Supplier: Ambeed
|
Warning
|
A1502989-100MG
A1502989-1G
A1502989-250MG
77875-822EA
339
USD
77875-822
77875-823
77875-824
2-(7,8-Difluoro-3-(methoxymethoxy)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(7,8-Difluoro-3-(methoxymethoxy)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₈H₂₁BF₂O₄ |
CAS Number:
2621935-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...