7-Fluoro-3-(methoxymethoxy)-8-((triisopropylsilyl)ethynyl)naphthalen-1-yl trifluoromethanesulfonate ≥97%

Supplier: Ambeed
Danger

A1502826-1G A1502826-100MG A1502826-250MG
77624-718EA 156.56 USD
77624-718 77875-814 77875-815
7-Fluoro-3-(methoxymethoxy)-8-((triisopropylsilyl)ethynyl)naphthalen-1-yl trifluoromethanesulfonate ≥97%
7-Fluoro-3-(methoxymethoxy)-8-((triisopropylsilyl)ethynyl)naphthalen-1-yl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₂₄H₃₀F₄O₅SSi
MDL Number: MFCD34471658
CAS Number: 2621932-36-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR