(11bS)-2,6-Bis(3,5-difluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%
Supplier: Ambeed
|
Warning
|
A1502341-100MG
A1502341-1G
A1502341-250MG
77597-774EA
138.38
USD
77597-774
77624-712
77624-714
(11bS)-2,6-Bis(3,5-difluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%
(11bS)-2,6-Bis(3,5-difluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
Precaución: for Research Use Only
Formula:
C₃₂H₁₇F₄O₄P MW: 572.44 g/mol |
CAS Number:
1620003-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...