4-Hexyl-4'-propyl-[1,1'-bi(cyclohexane)]-4-carbonitrile ≥95%

Supplier: Ambeed
Warning

A1502023-5G A1502023-100G A1502023-25G
77624-704EA 9.27 USD
77624-704 77875-784 77875-785
4-Hexyl-4'-propyl-[1,1'-bi(cyclohexane)]-4-carbonitrile ≥95%
4-Hexyl-4'-propyl-[1,1'-bi(cyclohexane)]-4-carbonitrile

Precaución: for Research Use Only

Formula: C₂₂H₃₉N
MW: 317.55 g/mol
CAS Number: 91174-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR