4-Pentyl-4'-(p-tolyl)-1,1'-bi(cyclohexane) ≥95%

Supplier: Ambeed
Warning

A1502021-100G A1502021-5G
77875-782EA 84 USD
77875-782 77875-783
4-Pentyl-4'-(p-tolyl)-1,1'-bi(cyclohexane) ≥95%
4-Pentyl-4'-(p-tolyl)-1,1'-bi(cyclohexane)

Precaución: for Research Use Only

Formula: C₂₄H₃₈
CAS Number: 2828433-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR