Copper(1+), bis(2,9-diphenyl-1,10-phenanthroline-κN1,κN10)-, (T-4)-, hexafluorophosphate(1-) (1:1) ≥97%

Supplier: Ambeed
Warning

A1501941-1G A1501941-250MG
77624-700EA 432.6 USD
77624-700 77624-702
Copper(1+), bis(2,9-diphenyl-1,10-phenanthroline-κN1,κN10)-, (T-4)-, hexafluorophosphate(1-) (1:1) ≥97%
Copper(1+), bis(2,9-diphenyl-1,10-phenanthroline-κN1,κN10)-, (T-4)-, hexafluorophosphate(1-) (1:1)

Precaución: for Research Use Only

Formula: C₄₈H₃₂CuF₆N₄P
MW: 873.3 g/mol
CAS Number: 183294-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR