N-[(11bS)-4-[[(11bS)-2,6-Diphenyl-4-[[(trifluoromethyl)sulfonyl]amino]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]amino]-2,6-diphenyl-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]-1,1,1-trifluoro-Methanesulfonamide ≥95%
Supplier: Ambeed
|
Warning
|
A1501502-100MG
A1501502-50MG
77592-824EA
218.33
USD
77592-824
77592-826
N-[(11bS)-4-[[(11bS)-2,6-Diphenyl-4-[[(trifluoromethyl)sulfonyl]amino]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]amino]-2,6-diphenyl-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]-1,1,1-trifluoro-Methanesulfonamide ≥95%
N-[(11bS)-4-[[(11bS)-2,6-Diphenyl-4-[[(trifluoromethyl)sulfonyl]amino]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]amino]-2,6-diphenyl-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]-1,1,1-trifluoro-Methanesulfonamide
Precaución: for Research Use Only
Formula:
C₆₆H₄₁F₆N₃O₈P₂S₂ MW: 1244.11 g/mol |
CAS Number:
2016814-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...