N-[(11bS)-4-[[(11bS)-2,6-Bis[3,5-bis(trifluoromethyl)phenyl]-4-[[(trifluoromethyl)sulfonyl]amino]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]amino]-2,6-bis[3,5-bis(trifluoromethyl)phenyl]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]-1,1,1-trifluoro-Methanesulfonamide ≥97%

Supplier: Ambeed
Warning

A1501494-100MG A1501494-250MG
77597-772EA 382.33 USD
77597-772 77624-690
N-[(11bS)-4-[[(11bS)-2,6-Bis[3,5-bis(trifluoromethyl)phenyl]-4-[[(trifluoromethyl)sulfonyl]amino]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]amino]-2,6-bis[3,5-bis(trifluoromethyl)phenyl]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]-1,1,1-trifluoro-Methanesulfonamide ≥97%
N-[(11bS)-4-[[(11bS)-2,6-Bis[3,5-bis(trifluoromethyl)phenyl]-4-[[(trifluoromethyl)sulfonyl]amino]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]amino]-2,6-bis[3,5-bis(trifluoromethyl)phenyl]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphep

Precaución: for Research Use Only

Formula: C₇₄H₃₃F₃₀N₃O₈P₂S₂
MW: 1788.1 g/mol
CAS Number: 2016814-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR