N-[(11bS)-4-[[(11bS)-2,6-Bis[4-(1,1-dimethylethyl)phenyl]-4-[[(trifluoromethyl)sulfonyl]amino]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]amino]-2,6-bis[4-(1,1-dimethylethyl)phenyl]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]-1,1,1-trifluoro-Methanesulfonamide ≥95%
Supplier: Ambeed
|
Warning
|
A1501492-100MG
A1501492-25MG
77624-686EA
313.12
USD
77624-686
77624-688
N-[(11bS)-4-[[(11bS)-2,6-Bis[4-(1,1-dimethylethyl)phenyl]-4-[[(trifluoromethyl)sulfonyl]amino]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]amino]-2,6-bis[4-(1,1-dimethylethyl)phenyl]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]-1,1,1-trifluoro-Methanesulfonamide ≥95%
N-[(11bS)-4-[[(11bS)-2,6-Bis[4-(1,1-dimethylethyl)phenyl]-4-[[(trifluoromethyl)sulfonyl]amino]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-ylidene]amino]-2,6-bis[4-(1,1-dimethylethyl)phenyl]-4λ5-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-y
Precaución: for Research Use Only
Formula:
C₈₂H₇₃F₆N₃O₈P₂S₂ MW: 1468.54 g/mol |
CAS Number:
2073168-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...