(4S,4'S)-4,4'-Di-tert-butyl-1,1'-di-p-tolyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%

Supplier: Ambeed
Warning

A1501182-100MG A1501182-250MG A1501182-1G
77597-768EA 149.65 USD
77597-768 77875-732 77875-731
(4S,4'S)-4,4'-Di-tert-butyl-1,1'-di-p-tolyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%
(4S,4'S)-4,4'-Di-tert-butyl-1,1'-di-p-tolyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole

Precaución: for Research Use Only

Formula: C₂₈H₃₈N₄
CAS Number: 1827669-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR